siRNA Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007983.2 | NP_032009.2 | 18, 19 | 2265 | ||
| GenBank mRNA | AK053752.1 | - | 18, 19 | 2200 | ||
| BC065098.1 | - | 18, 19 | 2265 | 30362116 | ||
| U39643.1 | - | 18, 19 | 2275 | |||

Faf1
Genomic Map