siRNA Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010176.4 | NP_034306.2 | 11 | 1166 | ||
| GenBank mRNA | AK143759.1 | - | 11 | 995 | ||
| AK158808.1 | - | 11 | 1052 | |||
| BC010767.1 | - | 11 | 951 | 4242421 | ||
| M84145.1 | - | 11 | 992 | |||
| Z11774.1 | - | 11 | 978 | |||

Fah
Genomic Map