siRNA Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016925.3 | NP_058621.2 | 11 | 973 | ||
| GenBank mRNA | AF178934.1 | - | 11 | 969 | ||
| AF208116.1 | - | 11 | 975 | |||
| AF208117.1 | - | 8 | 656 | |||
| AF208118.1 | - | 11 | 975 | |||
| AK084990.1 | - | 10 | 1081 | |||
| AK166112.1 | - | 11 | 958 | |||
| AK194298.1 | - | 1 | 46 | |||
| BC150705.1 | - | 11 | 971 | 9087616 | ||

Fanca
Genomic Map