siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 9 | 1214 | ||
| NM_001282942.1 | NP_001269871.1 | 8 | 1017 | |||
| NM_007985.3 | NP_032011.2 | 9 | 1214 | |||
| GenBank mRNA | AK035488.1 | - | 9 | 968 | ||
| AK042226.1 | - | 9 | 1184 | |||
| AK080697.1 | - | 1195 | ||||
| AK082717.1 | - | 8 | 995 | |||
| AK147025.1 | - | 9 | 1194 | |||
| AK163161.1 | - | 9 | 1215 | |||
| BC038609.1 | - | 8 | 999 | 5375091 | ||

Fancc
Genomic Map