siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 14 | 1675 | ||
| NM_001282942.1 | NP_001269871.1 | 13 | 1478 | |||
| NM_007985.3 | NP_032011.2 | 14 | 1675 | |||
| GenBank mRNA | AK035488.1 | - | 14 | 1429 | ||
| AK042226.1 | - | 14 | 1645 | |||
| AK082717.1 | - | 11 | 1196 | |||
| AK147025.1 | - | 14 | 1655 | |||
| BC038609.1 | - | 13 | 1460 | 5375091 | ||
| L08266.1 | - | 14 | 1656 | |||

Fancc
Genomic Map