siRNA Details
| Gene Symbol: Fap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007986.3 | NP_032012.1 | 20 | 1933 | ||
| GenBank mRNA | BC019190.1 | - | 20 | 1837 | 5028579 | |
| Y10007.1 | - | 20 | 1933 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fap| Gene Symbol: Fap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007986.3 | NP_032012.1 | 20 | 1933 | ||
| GenBank mRNA | BC019190.1 | - | 20 | 1837 | 5028579 | |
| Y10007.1 | - | 20 | 1933 | |||