siRNA Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007987.2 | NP_032013.2 | 6 | 569 | ||
| GenBank mRNA | AK002590.1 | - | 6 | 520 | ||
| BC061160.1 | - | 6 | 542 | 30302649 | ||
| DQ846748.1 | - | 6 | 513 | |||
| M83649.1 | - | 6 | 562 | |||

Fas
Genomic Map