siRNA Details
| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007988.3 | NP_032014.3 | 6 | 1223 | ||
| GenBank mRNA | AF127033.1 | - | 6 | 740 | ||
| AK147214.1 | - | 6 | 919 | |||
| AK147694.1 | - | 6 | 986 | |||
| AK171499.1 | - | 6 | 855 | |||
| BC046513.1 | - | 6 | 777 | 4196969 | ||

Fasn
Genomic Map