siRNA Details
| Gene Symbol: Ms4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276328.1 | NP_001263257.1 | 3 | 350 | ||
| NM_001276330.1 | NP_001263259.1 | 3 | 350 | |||
| NM_013516.2 | NP_038544.1 | 3 | 350 | |||
| GenBank mRNA | AK144021.1 | - | 3 | 351 | ||
| BB847820.1 | - | 3 | 351 | |||
| BC139323.1 | - | 3 | 274 | 8862345 | ||
| BC139324.1 | - | 3 | 274 | 8862346 | ||
| CN663232.1 | - | 3 | 290 | |||
| J05019.1 | - | 3 | 274 | |||

Ms4a2
Genomic Map