siRNA Details
| Gene Symbol: Fcgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010186.5 | NP_034316.1 | 5 | 669 | ||
| GenBank mRNA | AF143154.1 | - | 5 | 662 | ||
| AF143155.1 | - | 5 | 662 | |||
| AF143156.1 | - | 5 | 662 | |||
| AF143157.1 | - | 5 | 662 | |||
| AF143158.1 | - | 5 | 662 | |||
| AF143159.1 | - | 5 | 662 | |||
| AF143160.1 | - | 5 | 662 | |||
| AF143161.1 | - | 5 | 662 | |||
| AF143162.1 | - | 5 | 662 | |||
| AF143163.1 | - | 5 | 662 | |||
| AF143164.1 | - | 5 | 662 | |||
| AF143165.1 | - | 5 | 662 | |||
| AF143166.1 | - | 5 | 662 | |||
| AF143167.1 | - | 5 | 662 | |||
| AF143168.1 | - | 5 | 662 | |||
| AF143169.1 | - | 5 | 662 | |||
| AF143170.1 | - | 5 | 662 | |||
| AF143171.1 | - | 5 | 662 | |||
| AF143172.1 | - | 5 | 662 | |||
| AF143173.1 | - | 5 | 662 | |||
| AF143174.1 | - | 5 | 662 | |||
| AF143175.1 | - | 5 | 662 | |||
| AF143176.1 | - | 5 | 662 | |||
| AF143177.1 | - | 5 | 662 | |||
| AF143178.1 | - | 5 | 662 | |||
| AF143179.1 | - | 5 | 662 | |||
| AF143180.1 | - | 5 | 662 | |||
| AF143181.1 | - | 5 | 677 | |||
| AF143186.1 | - | 5 | 662 | |||
| AF143187.1 | - | 5 | 662 | |||
| AF143188.1 | - | 5 | 662 | |||
| AK033874.1 | - | 5 | 670 | |||
| BC025535.1 | - | 5 | 659 | |||
| M31314.1 | - | 5 | 663 | |||
| X70980.1 | - | 5 | 677 | |||

Fcgr1
Genomic Map