siRNA Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007995.3 | NP_032021.1 | 9 | 776 | ||
| GenBank mRNA | AB007813.1 | - | 9 | 668 | ||
| AB222271.1 | - | 8 | 557 | |||
| AK133539.1 | - | 9 | 753 | |||
| AK138752.1 | - | 9 | 723 | |||
| AK159651.1 | - | 9 | 746 | |||
| BC019180.1 | - | 9 | 724 | 5008934 | ||
| CT010383.1 | - | 9 | 638 | |||

Fcna
Genomic Map