siRNA Details
| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277268.1 | NP_001264197.1 | 2 | 625 | ||
| NM_010203.5 | NP_034333.1 | 3 | 729 | |||
| GenBank mRNA | AB016516.1 | - | 2 | 397 | ||
| AK028694.1 | - | 3 | 586 | |||
| AK028894.1 | - | 3 | 731 | |||
| BC071227.1 | - | 3 | 702 | 30443152 | ||
| M30643.1 | - | 3 | 501 | |||

Fgf5
Genomic Map