siRNA Details
| Gene Symbol: Fgfr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008011.2 | NP_032037.2 | 12 | 1728 | ||
| GenBank mRNA | AK084850.1 | - | 12 | 1728 | ||
| AK085723.1 | - | 12 | 1726 | |||
| AY493377.2 | - | 11 | 1614 | |||
| BC033313.1 | - | 12 | 1915 | 5132831 | ||
| DQ388428.1 | - | 11 | 1614 | |||
| X59927.1 | - | 12 | 1697 | |||

Fgfr4
Genomic Map