siRNA Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289533.1 | NP_001276462.1 | 5 | 651 | ||
| NM_010212.4 | NP_034342.1 | 5 | 800 | |||
| GenBank mRNA | AF055889.1 | - | 4 | 528 | ||
| AF114381.1 | - | 4 | 528 | |||
| AF153340.1 | - | 4 | 572 | |||
| AK052936.1 | - | 5 | 652 | |||
| AK133708.1 | - | 5 | 772 | |||
| AK147024.1 | - | 5 | 801 | |||
| BC021468.1 | - | 5 | 709 | 5066565 | ||
| U77040.2 | - | 5 | 690 | |||

Fhl2
Genomic Map