siRNA Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289533.1 | NP_001276462.1 | 3 | 412 | ||
| NM_010212.4 | NP_034342.1 | 3 | 561 | |||
| GenBank mRNA | AF153340.1 | - | 2 | 333 | ||
| AK052936.1 | - | 3 | 413 | |||
| AK133708.1 | - | 3 | 534 | |||
| AK147024.1 | - | 3 | 562 | |||
| BC021468.1 | - | 3 | 470 | 5066565 | ||
| U77040.2 | - | 3 | 451 | |||

Fhl2
Genomic Map