siRNA Details
| Gene Symbol: Smc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301412.1 | NP_001288341.1 | 10 | 1410 | ||
| NM_008017.4 | NP_032043.3 | 10 | 1334 | |||
| GenBank mRNA | AJ534939.1 | - | 9 | 1165 | ||
| AK017747.1 | - | 10 | 1312 | |||
| AK145341.1 | - | 10 | 1364 | |||
| AK152090.1 | - | 10 | 1362 | |||
| BC011064.1 | - | 10 | 1272 | |||
| BC021847.1 | - | 10 | 1424 | |||
| BC094380.1 | - | 10 | 1306 | 30543190 | ||

Smc2
Genomic Map