siRNA Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164717.1 | NP_001158189.1 | 11 | 1210 | ||
| NM_008018.4 | NP_032044.2 | 12 | 1294 | |||
| GenBank mRNA | AJ007012.1 | - | 12 | 1294 | ||
| AK085227.1 | - | 5 | 1133 | |||
| BC118022.1 | - | 11 | 1046 | 40097448 | ||

Sh3pxd2a
Genomic Map