siRNA Details
| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010228.3 | NP_034358.2 | 16, 17 | 2614 | ||
| GenBank mRNA | D88689.1 | - | 16, 17 | 2611 | ||
| L07297.1 | - | 16, 17 | 2386 | |||
| X78568.1 | - | 16, 17 | 2599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flt1| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010228.3 | NP_034358.2 | 16, 17 | 2614 | ||
| GenBank mRNA | D88689.1 | - | 16, 17 | 2611 | ||
| L07297.1 | - | 16, 17 | 2386 | |||
| X78568.1 | - | 16, 17 | 2599 | |||