siRNA Details
| Gene Symbol: Flt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010229.2 | NP_034359.2 | 9 | 1313 | ||
| GenBank mRNA | AK045865.1 | - | 9 | 1136 | ||
| AK149292.1 | - | 9 | 1131 | |||
| AK163640.1 | - | 9 | 1314 | |||
| BC109003.1 | - | 9 | 1106 | |||
| BC109004.1 | - | 9 | 1118 | |||
| M64689.1 | - | 9 | 1119 | |||
| X59398.1 | - | 9 | 1170 | |||

Flt3
Genomic Map