siRNA Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013520.3 | NP_038548.3 | 7 | 611 | ||
| GenBank mRNA | AK020105.1 | - | 7 | 611 | ||
| BB224229.2 | - | 1 | 95 | |||
| BC019801.1 | - | 7 | 580 | 5132319 | ||
| CK622267.1 | - | 7 | 581 | |||
| EU274583.1 | - | 6 | 523 | |||
| L23636.1 | - | 6 | 554 | |||
| U04807.1 | - | 6 | 778 | |||

Flt3l
Genomic Map