siRNA Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013520.3 | NP_038548.3 | 4 | 248 | ||
| GenBank mRNA | AK020105.1 | - | 4 | 248 | ||
| BC019801.1 | - | 4 | 233 | 5132319 | ||
| CK622267.1 | - | 4 | 218 | |||
| EU274583.1 | - | 3 | 160 | |||
| L23636.1 | - | 3 | 191 | |||
| S76459.1 | - | 3 | 166 | |||
| U04807.1 | - | 3 | 415 | |||

Flt3l
Genomic Map