siRNA Details
| Gene Symbol: Fmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285458.1 | NP_001272387.1 | 9 | 3363 | ||
| NM_001285459.1 | NP_001272388.1 | 9 | 2987 | |||
| NM_010230.2 | NP_034360.2 | 11 | 3657 | |||
| GenBank mRNA | BC138036.1 | - | 8 | 3269 | 8861055 | |
| BC171945.1 | - | 8 | 3263 | 9054231 | ||
| CK391697.1 | - | 5 | 289 | |||
| CO814739.2 | - | 4 | 214 | |||
| X53599.1 | - | 11 | 3617 | |||
| X62379.1 | - | 9 | 2993 | |||

Fmn1
Genomic Map