siRNA Details
| Gene Symbol: Fmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285458.1 | NP_001272387.1 | 2 | 268 | ||
| NM_010230.2 | NP_034360.2 | 2 | 268 | |||
| GenBank mRNA | AK135758.1 | - | 2 | 234 | ||
| BC138036.1 | - | 1 | 168 | 8861055 | ||
| BC171945.1 | - | 1 | 168 | 9054231 | ||
| BY288387.1 | - | 2 | 269 | |||
| X53599.1 | - | 2 | 222 | |||

Fmn1
Genomic Map