siRNA Details
| Gene Symbol: Fos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010234.2 | NP_034364.1 | 3 | 559 | ||
| GenBank mRNA | AK154418.1 | - | 3 | 571 | ||
| BC029814.1 | - | 3 | 559 | 2582234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fos| Gene Symbol: Fos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010234.2 | NP_034364.1 | 3 | 559 | ||
| GenBank mRNA | AK154418.1 | - | 3 | 571 | ||
| BC029814.1 | - | 3 | 559 | 2582234 | ||