siRNA Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080845.2 | NP_543121.1 | 6 | 733 | ||
| GenBank mRNA | AK165414.1 | - | 5 | 642 | ||
| BC010813.1 | - | 6 | 695 | 4211065 | ||
| BC024078.1 | - | 6 | 727 | 5098529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ftcd| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080845.2 | NP_543121.1 | 6 | 733 | ||
| GenBank mRNA | AK165414.1 | - | 5 | 642 | ||
| BC010813.1 | - | 6 | 695 | 4211065 | ||
| BC024078.1 | - | 6 | 727 | 5098529 | ||