siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017861.1 | - | 4 | 455 | ||
| AK146459.1 | - | 5 | 571 | |||
| AK158704.1 | - | 5 | 573 | |||
| BC008130.1 | - | 6 | 576 | 3498984 | ||
| BC034861.1 | - | 5 | 580 | |||
| X71978.1 | - | 5 | 595 | |||
| Z67963.1 | - | 5 | 595 | |||

Aktip
Genomic Map