siRNA Details
| Gene Symbol: Fut4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010242.3 | NP_034372.1 | 1 | 1258 | ||
| GenBank mRNA | AK041752.1 | - | 1 | 1258 | ||
| BC137588.1 | - | 1 | 1249 | 8860607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut4| Gene Symbol: Fut4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010242.3 | NP_034372.1 | 1 | 1258 | ||
| GenBank mRNA | AK041752.1 | - | 1 | 1258 | ||
| BC137588.1 | - | 1 | 1249 | 8860607 | ||