siRNA Details
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113188.1 | NP_001106659.1 | 7 | 619 | ||
| NM_001113189.1 | NP_001106660.1 | 7 | 619 | |||
| NM_008053.2 | NP_032079.1 | 7 | 619 | |||
| GenBank mRNA | AF124385.1 | - | 2 | 120 | ||
| BC019139.1 | - | 7 | 600 | 5041635 | ||
| X90875.1 | - | 7 | 598 | |||

Fxr1
Genomic Map