siRNA Details
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113188.1 | NP_001106659.1 | 14 | 1687 | ||
| NM_001113189.1 | NP_001106660.1 | 15 | 1600 | |||
| NM_008053.2 | NP_032079.1 | 15 | 1600 | |||
| GenBank mRNA | AF124385.1 | - | 10 | 1101 | ||
| AK049231.1 | - | 6 | 650 | |||
| AK178268.1 | - | 1 | 54 | |||
| BC019139.1 | - | 15 | 1581 | 5041635 | ||
| CB196254.1 | - | 2 | 223 | |||
| CF170707.1 | - | 2 | 265 | |||
| X90875.1 | - | 15 | 1579 | |||

Fxr1
Genomic Map