siRNA Details
| Gene Symbol: Fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162494.1 | NP_001155966.1 | 3 | 509 | ||
| NM_008056.3 | NP_032082.3 | 3 | 513 | |||
| GenBank mRNA | AK088399.1 | - | 3 | 378 | ||
| BC026150.1 | - | 3 | 502 | 3983985 | ||
| CD348842.1 | - | 3 | 479 | |||
| U43319.1 | - | 2 | 339 | |||
More Information
Set Membership: |
|

Fzd6
Genomic Map