siRNA Details
Gene Symbol: Fzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008057.3 | NP_032083.3 | 1 | 1839 | ||
GenBank mRNA | BC049781.1 | - | 1 | 1562 | ||
BC063077.1 | - | 2 | 1170 | 6844727 | ||
U43320.1 | - | 1 | 1387 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
