siRNA Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010247.2 | NP_034377.2 | 7 | 912 | ||
| GenBank mRNA | AB010282.1 | - | 7 | 853 | ||
| AF483500.1 | - | 6 | 792 | |||
| AF483501.1 | - | 6 | 792 | |||
| AK143570.1 | - | 7 | 887 | |||
| AK146392.1 | - | 7 | 831 | |||
| AK146394.1 | - | 7 | 831 | |||
| AK146396.1 | - | 7 | 879 | |||
| BC031422.1 | - | 7 | 861 | 3669715 | ||
| BC051085.1 | - | 7 | 2090 | |||
| M38700.1 | - | 6 | 754 | |||

Xrcc6
Genomic Map