siRNA Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010247.2 | NP_034377.2 | 7 | 1021 | ||
| GenBank mRNA | AB010282.1 | - | 7 | 962 | ||
| AF483500.1 | - | 6 | 901 | |||
| AF483501.1 | - | 6 | 901 | |||
| AK143570.1 | - | 7 | 996 | |||
| AK146392.1 | - | 7 | 940 | |||
| AK146394.1 | - | 7 | 940 | |||
| AK146396.1 | - | 7 | 988 | |||
| BC031422.1 | - | 7 | 970 | 3669715 | ||
| BC051085.1 | - | 7 | 2199 | |||
| M38700.1 | - | 6 | 863 | |||

Xrcc6
Genomic Map