siRNA Details
| Gene Symbol: Gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301298.1 | NP_001288227.1 | 4 | 2073 | ||
| NM_021356.2 | NP_067331.2 | 4 | 2033 | |||
| GenBank mRNA | AJ250669.1 | - | 4 | 1180 | ||
| AK132176.1 | - | 4 | 2074 | |||
| AK155423.1 | - | 4 | 2044 | |||
| BC007483.1 | - | 4 | 2033 | 3495926 | ||
| BC094659.1 | - | 4 | 1384 | 6854020 | ||
| CT010264.1 | - | 4 | 1157 | |||

Gab1
Genomic Map