siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008066.3 | NP_032092.1 | 9 | 1250 | ||
| GenBank mRNA | AK039055.1 | - | 9 | 1251 | ||
| AK046953.1 | - | 4 | 1384 | |||
| AK048165.1 | - | 1148 | ||||
| BC115727.1 | - | 9 | 1172 | 40039253 | ||
| M86567.1 | - | 9 | 1001 | |||

Gabra2
Genomic Map