siRNA Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008067.4 | NP_032093.3 | 10 | 1482 | ||
| GenBank mRNA | AK039144.1 | - | 10 | 1423 | ||
| AK134760.1 | - | 10 | 1391 | |||
| BC119317.1 | - | 10 | 1280 | 8734066 | ||
| BC120686.1 | - | 10 | 1280 | 40129609 | ||
| CD354278.1 | - | 2 | 208 | |||
| M86568.1 | - | 10 | 1384 | |||

Gabra3
Genomic Map