siRNA Details
| Gene Symbol: Gabra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099641.2 | NP_001093111.1 | 4 | 721 | ||
| NM_008068.3 | NP_032094.2 | 4 | 691 | |||
| GenBank mRNA | AF256197.1 | - | 4 | 417 | ||
| AF256198.1 | - | 4 | 417 | |||
| AK135145.1 | - | 4 | 701 | |||
| AK137331.1 | - | 4 | 702 | |||
| BC145158.1 | - | 4 | 439 | 9053709 | ||
| BC145702.1 | - | 4 | 439 | 40131005 | ||
| X51986.1 | - | 4 | 627 | |||

Gabra6
Genomic Map