siRNA Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008078.2 | NP_032104.2 | 5 | 902 | ||
| GenBank mRNA | AF326549.1 | - | 5 | 566 | ||
| AF326550.1 | - | 5 | 566 | |||
| AK136834.1 | - | 5 | 602 | |||
| BC018380.1 | - | 5 | 617 | 4482097 | ||
| D42051.1 | - | 5 | 918 | |||
| KC134211.1 | - | 5 | 566 | |||
| L16980.1 | - | 5 | 638 | |||
| S67454.1 | - | 1 | 44 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012563, M72422.

Gad2
Genomic Map