siRNA Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015736.2 | NP_056551.2 | 7 | 1714 | ||
| GenBank mRNA | AK019995.1 | - | 7 | 1713 | ||
| AK135489.1 | - | 7 | 1675 | |||
| BC043331.1 | - | 7 | 1570 | 5342768 | ||
| U70538.1 | - | 7 | 1558 | |||

Galnt3
Genomic Map