siRNA Details
| Gene Symbol: Galr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010254.4 | NP_034384.3 | 2 | 1379 | ||
| GenBank mRNA | AF077375.1 | - | 2 | 829 | ||
| AK053776.1 | - | 2 | 1380 | |||
| AK132050.1 | - | 2 | 1187 | |||
| BC116980.1 | - | 2 | 1336 | 40126299 | ||
| BC116982.1 | - | 2 | 1336 | 40126301 | ||
More Information
Set Membership: |
|

Galr2
Genomic Map