siRNA Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088379.1 | - | 9 | 902 | ||
| AK134525.1 | - | 6 | 556 | |||
| AK163544.1 | - | 6 | 588 | |||
| AV027352.2 | - | 3 | 340 | |||
| BC010985.1 | - | 9 | 907 | 4038194 | ||
| M96265.1 | - | 9 | 931 | |||

Galt
Genomic Map