siRNA Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010256.2 | NP_034386.2 | 19, 20 | 2718 | ||
| GenBank mRNA | AK146355.1 | - | 19, 20 | 2703 | ||
| AK146478.1 | - | 19, 20 | 2660 | |||
| AK168458.1 | - | 19, 20 | 2679 | |||
| AK168501.1 | - | 19, 20 | 2681 | |||
| AK168724.1 | - | 19, 20 | 2693 | |||
| AK168796.1 | - | 19, 20 | 2693 | |||
| AK168864.1 | - | 19, 20 | 2669 | |||
| AK168876.1 | - | 19, 20 | 2670 | |||
| BC070465.1 | - | 19, 20 | 2665 | 30611835 | ||
| U01024.1 | - | 18, 19 | 2623 | |||

Gart
Genomic Map