siRNA Details
| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 5 | 1283 | ||
| GenBank mRNA | AK132020.1 | - | 5 | 1304 | ||
| AK142213.1 | - | 5 | 1285 | |||
| BC105653.1 | - | 6 | 996 | 40046623 | ||
| BC105654.1 | - | 6 | 1057 | 40046622 | ||
| BC105655.1 | - | 6 | 906 | 40046621 | ||
| BC105656.1 | - | 6 | 691 | |||
| U84725.1 | - | 5 | 1277 | |||

Gata5
Genomic Map