siRNA Details
| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 6 | 1421 | ||
| GenBank mRNA | AK132020.1 | - | 6 | 1442 | ||
| AK142213.1 | - | 6 | 1423 | |||
| BC105653.1 | - | 7 | 1134 | 40046623 | ||
| BC105654.1 | - | 7 | 1195 | 40046622 | ||
| BC105655.1 | - | 7 | 1044 | 40046621 | ||
| BC105656.1 | - | 7 | 829 | |||
| U84725.1 | - | 6 | 1415 | |||

Gata5
Genomic Map