siRNA Details
| Gene Symbol: Gba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077411.2 | NP_001070879.1 | 5 | 628 | ||
| NM_008094.5 | NP_032120.1 | 6 | 797 | |||
| GenBank mRNA | AK082767.1 | - | 5 | 626 | ||
| AK152745.1 | - | 5 | 610 | |||
| AK166198.1 | - | 6 | 788 | |||
| BC006663.1 | - | 6 | 782 | 3498551 | ||
| M24119.1 | - | 5 | 537 | |||

Gba
Genomic Map