siRNA Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301228.1 | NP_001288157.1 | 3 | 415 | ||
| NM_001301229.1 | NP_001288158.1 | 3 | 306 | |||
| NM_001301231.1 | NP_001288160.1 | 3 | 345 | |||
| NM_005342.3 | NP_005333.2 | 3 | 310 | |||
| GenBank mRNA | BC037210.1 | - | 2 | 215 | ||
| BC070482.1 | - | 4 | 394 | 6140626 | ||
| BF979310.1 | - | 3 | 351 | |||
| BX537505.1 | - | 3 | 295 | |||
| DB191695.1 | - | 3 | 295 | |||
| DN996967.1 | - | 3 | 317 | |||
| L05085.1 | - | 2 | 215 | |||
| Y10043.1 | - | 3 | 289 | |||

HMGB3
Genomic Map