siRNA Details
| Gene Symbol: Gcsam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159297.1 | NP_001152769.1 | 3, 4 | 255 | ||
| NM_008099.4 | NP_032125.3 | 2, 3 | 189 | |||
| GenBank mRNA | AK036553.1 | - | 2, 3 | 207 | ||
| AY500830.1 | - | 3, 4 | 163 | |||
| BC099585.1 | - | 2, 3 | 153 | 30289367 | ||
| U13263.1 | - | 2, 3 | 190 | |||

Gcsam
Genomic Map