siRNA Details
| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF276754.1 | - | 3 | 336 | ||
| AK007911.1 | - | 3 | 329 | |||
| AK148544.1 | - | 3 | 330 | |||
| BC012975.1 | - | 3 | 331 | 4218718 | ||
| Z46845.1 | - | 2 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcg| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF276754.1 | - | 3 | 336 | ||
| AK007911.1 | - | 3 | 329 | |||
| AK148544.1 | - | 3 | 330 | |||
| BC012975.1 | - | 3 | 331 | 4218718 | ||
| Z46845.1 | - | 2 | 264 | |||