siRNA Details
| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008103.3 | NP_032129.2 | 6 | 1422 | ||
| GenBank mRNA | AK142574.1 | - | 6 | 1408 | ||
| BC066866.1 | - | 6 | 1422 | 30432193 | ||
| U59876.1 | - | 6 | 1438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcm1| Gene Symbol: Gcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008103.3 | NP_032129.2 | 6 | 1422 | ||
| GenBank mRNA | AK142574.1 | - | 6 | 1408 | ||
| BC066866.1 | - | 6 | 1422 | 30432193 | ||
| U59876.1 | - | 6 | 1438 | |||