siRNA Details
| Gene Symbol: Nr6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159548.1 | NP_001153020.1 | 5 | 792 | ||
| NM_001159549.1 | NP_001153021.1 | 5 | 789 | |||
| NM_010264.4 | NP_034394.1 | 7 | 1100 | |||
| GenBank mRNA | AF390896.1 | - | 8 | 1245 | ||
| CK638822.1 | - | 1 | 189 | |||
| U09563.1 | - | 7 | 1100 | |||
| U14666.1 | - | 7 | 1059 | |||

Nr6a1
Genomic Map